Learn More
Thermo Scientific™ NUCLEIC-CARD™ COLOR matrix, 1 spot
NUCLEIC-CARD COLOR is an easy-to-use system for collection, preservation, and long-term storage of nucleic acids at room temperature.
Marca: Thermo Scientific™ 4473974
Descripción
The NUCLEIC-CARD COLOR matrix is chemically-treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature. NUCLEIC-CARD COLOR changes color upon sample application to facilitate processing of buccal samples. NUCLEIC-CARD collection devices are manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
Key Features:
• Enables direct PCR amplification from a card punch, eliminating extraction steps
• Facilitates high quality STR profiles with Applied Biosystems direct PCR amplification kits
• Certified free of DNase, Rnase, and amplifiable human DNA
• Prevents growth of microorganisms to enable long-term room-temperature storage
• Easily automated with commercially-available punching systems to minimize manual handling and maximize sample integrity
For Forensic or Paternity Use Only.
NUCLEIC-CARD is a trademark of Copan Flock Technologies.
Especificaciones
50 NUCLEIC-CARD COLOR 1 Spot Store (at room temperature) |
|
Harris Manual Punching System, BSD Robotics Punching System | |
50 Cards |
Card | |
NUCLEIC-CARD | |
Temperatura ambiente |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.