Learn More
Thermo Scientific™ NUCLEIC-CARD™ COLOR matrix, 1 spot
NUCLEIC-CARD COLOR is an easy-to-use system for collection, preservation, and long-term storage of nucleic acids at room temperature.
Marca:  Thermo Scientific™ 4473974
Código de producto. 15662406
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Descripción
Descripción
The NUCLEIC-CARD COLOR matrix is chemically-treated to enable cell lysis and protein denaturation. Nucleic acids are immobilized and preserved for long-term storage at room temperature. NUCLEIC-CARD COLOR changes color upon sample application to facilitate processing of buccal samples. NUCLEIC-CARD collection devices are manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
Key Features:
• Enables direct PCR amplification from a card punch, eliminating extraction steps
• Facilitates high quality STR profiles with Applied Biosystems direct PCR amplification kits
• Certified free of DNase, Rnase, and amplifiable human DNA
• Prevents growth of microorganisms to enable long-term room-temperature storage
• Easily automated with commercially-available punching systems to minimize manual handling and maximize sample integrity
For Forensic or Paternity Use Only.
NUCLEIC-CARD is a trademark of Copan Flock Technologies.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
50 NUCLEIC-CARD COLOR 1 Spot
Store (at room temperature)
Harris Manual Punching System, BSD Robotics Punching System
50 Cards
Card
NUCLEIC-CARD
Temperatura ambiente
SDS
Documentos Certificados
Promociones
Promociones
Título del producto
Thermo Scientific™ NUCLEIC-CARD™ COLOR matrix, 1 spot > 50 NUCLEIC-CARD COLOR 1 Spot