missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NPFFR2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand: Abnova™ H00010886-T01.100uL
This item is not returnable.
View return policy
Specifications
293T | |
NPFFR2 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 48.7kDa | |
Transient overexpression cell lysate was tested with anti-NRG4antibody (Mfr. No. H00010886-B01P) by Western Blot. | |
Tampón de muestra 1X (Tris-HCl 50 mM, SDS al 2 %, glicerol al 10 %, 2-mercaptoetanol 300 mM, azul de bromofenol al 0,01 %) | |
Almacenar a -80°C. Dividir para evitar repetir la congelación y descongelación. |
Western Blot | |
NM_053036 | |
pCMV-NPFFR2 full-length | |
100 μl | |
Human |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UL NPFFR2 293T Cell Transient Overexpression Ly
Signal Word
- Atención
Hazard Category
- Sensibilización cutánea Categoría 1
Hazard Statement
- H317-Puede provocar una reacción alérgica en la piel.
Precautionary Statement
- P261-Evitar respirar el polvo/el humo/el gas/la niebla/los vapores/el aerosol.
- P272-Las prendas de trabajo contaminadas no podrán sacarse del lugar de trabajo.
- P280-Llevar guantes/prendas/gafas/máscara de protección.
Supplemental information
- MIXTURE LIST-Contém: Mercaptoethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ NPFFR2 293T Cell Transient Overexpression Lysate (Denatured) > 100 μl
Spot an opportunity for improvement?Share a Content Correction