missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Noradrenaline Competitive ELISA Kit
ELISA
Marca: Invitrogen™ EEL010
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Norepinephrine (NE), also called noradrenalin (NA), is an organic chemical in the catecholamine family that functions in the brain and body as both a hormone and neurotransmitter. NA/NE acts on target cells by binding and activating adrenergic receptors located on the cell surface to mobilize the brain and body for action. Its differs structurally from epinephrine in that the methyl group attached to its nitrogen is replaced by a hydrogen atom in NE.
Especificaciones
De 0,31 a 20 ng/ml | |
HRP | |
Plasma, Serum | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 pruebas | |
Plasma 50 μL, Serum 50 μL | |
Noradrenaline | |
3 h 30 min |
0.19 ng/mL | |
ELISA | |
Lector de microplacas colorimétrico | |
<10 | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 h 20 min |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ Noradrenaline Competitive ELISA Kit > 96 pruebas
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido