Learn More
NMDAR2B Monoclonal Antibody (NR2B), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA12014
Descripción
MA1-2014 detects the N-methyl-D-aspartate (NMDA) receptor type 2B in human and mouse samples. MA1-2014 has been used successfully in Western blot, IHC (P) and immunocytochemistry procedures. In Western blot analysis of mouse brain tissue this antibody detects a ∽166 kDa protein representing NMDA receptor type 2B. The MA1-2014 immunogen is a recombinant protein composed of amino acid residues 934-1457 of the rat NMDA receptor type 2B.
NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer’s Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal dominant 6-related mental retardation.
Especificaciones
NMDAR2B | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q00960, Q01097, Q13224 | |
GRIN2B | |
Recombinant protein: NR2B subunit of amino acid residues 934-1457. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
NR2B | |
Unconjugated | |
GRIN2B | |
AW490526; EIEE27; GluN2B; GluRepsilon2; glutamate [NMDA] receptor subunit epsilon-2; glutamate ionotropic receptor NMDA type subunit 2B; glutamate receptor; glutamate receptor ionotropic, NMDA 2B; glutamate receptor subunit epsilon-2; glutamate receptor, ionotropic, NMDA2B; glutamate receptor, ionotropic, NMDA2B (epsilon 2); glutamate receptor, ionotropic, N-methyl D-aspartate 2B; GRIN 2B; GRIN2B; HGNC:4586; hNR 3; hNR3; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2B; MGC142178; MGC142180; MRD6; Nmdar2b; NMDE2; NME2; N-methyl D-aspartate receptor 2B; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 2B; N-methyl-D-aspartate receptor subunit 3; NR2B; NR3 | |
Mouse | |
Protein A | |
RUO | |
14812, 24410, 2904 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
Número de lote | Tipo de certificado | Fecha | Catalog Number |
---|---|---|---|
AC409170 | Certificado de análisis | 24/03/2025 |
Promociones
-
Multiple Manufacturers
Impulse la proteómica
Desbloquee avances en proteómica y ahorre de manera exclusiva.Promo Code: 25977Expires: 2025-05-31
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.