missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2B Antibody (NR2B), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Marca: Novus Biologicals NB100-74475
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NMDAR2B Monoclonal specifically detects NMDAR2B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
NMDAR2B | |
Monoclonal | |
1 mg/mL | |
Western Blot 2 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10 - 1:100, Immunocytochemistry 1 ug/ml | |
glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3 | |
Mouse | |
166 kDa | |
100 μg | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
2904 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
NR2B | |
Unconjugated | |
Q13224 | |
GRIN2B | |
Recombinant protein: NR2B subunit of amino acid residues 934-1457. | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
NMDAR2B Antibody (NR2B), Novus Biologicals™ > 100 ug; Unconjugated
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido