Learn More
Invitrogen™ Human NIM1, GST Tag Recombinant Protein
NIM1K Recombinant Human Protein or NIM1 belongs to the CAMK kinase family.
Marca:  Invitrogen™ A33363
Detalles adicionales : Peso : 0.00900kg
 Ver más versiones de este producto
Código de producto. 17198775
  • 7580.00€ / 1 miligramo
Fecha estimada de envío: 03-06-2024
para ver el stock
Descripción
Descripción
Unlike to other AMPK-related kinases, NIM1 cannot be phosphorylated and activated by LKB1. Two somatic mutations (P333S and P411T) were found in lung neuroendocrine carcinoma and lung large cell carcinoma samples.
  • Validated for use with the Z’-LYTE™ assay
  • Available in multiple pack sizes for convenient use
Especificaciones
Especificaciones
NP_699192
75.9 kDa
CAMK Unique Kinase Family
Human
GST Tag
Dry Ice
Full Length, Recombinant, Active
Kinase Assay
1 mg
Full Length
Recombinant
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Insect