missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NIM1, GST Tag Recombinant Protein
NIM1K Recombinant Human Protein or NIM1 belongs to the CAMK kinase family.
Marca: Invitrogen™ A32873
Detalles adicionales : Peso : 0.00900kg
Descripción
Unlike to other AMPK-related kinases, NIM1 cannot be phosphorylated and activated by LKB1. Two somatic mutations (P333S and P411T) were found in lung neuroendocrine carcinoma and lung large cell carcinoma samples.- Validated for use with the Z’-LYTE™ assay
- Available in multiple pack sizes for convenient use
Especificaciones
NP_699192 | |
75.9 kDa | |
CAMK Unique Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant, Active |
Kinase Assay | |
100 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid |