Learn More
NGDN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA551489
Descripción
Immunogen sequence: LMYLMDLTHL ILDKASGGSL QGHDAVLRLV EIRTVLEKLR PLDQKLKYQI DKLIKTAVTG SLSENDPLRF KPHPSNMMSK LSSEDEEEDE AEDDQSEASG KKSVKGVSKK YVPPRLVPVH YDETEAEREK KRLERAKRRA LSSSVIREL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 95%.
Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
Especificaciones
NGDN | |
Polyclonal | |
Unconjugated | |
Ngdn | |
1500001L15Rik; C14orf120; CANu1; Centromere accumulated nuclear protein 1; EIF4E-binding protein; eukaryotic initiation factor 4E and CPEB binding protein; LCP5; lpd-2; neuroguidin; neuroguidin, EIF4E binding protein; NGD; Ngdn; RGD1310211 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25983 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NEJ9 | |
Ngdn | |
Recombinant protein corresponding to Human NGDN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Research Use Only