missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human NEK2, His Tag Recombinant Protein
Marca: Thermo Scientific™ PV4026
Detalles adicionales : Peso : 0.00900kg
Descripción
- NEK2 is involved in cell cycle regulation
- It is essential for faithful chromosome segregation
Especificaciones
NP_002488 | |
54.5 kDa | |
NEK2, NLK1 | |
Human | |
His Tag | |
Ser/Thr Kinases: Other | |
Baculovirus | |
Frozen |
Kinase Assay | |
1 mg | |
NEK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 500mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Full Length | |
Drug Development |