missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NBL1 (Human) Recombinant Protein
Human NBL1 full-length ORF ( AAH12037, 1 a.a. - 180 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00004681-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MLRVLVGAVLPAMLLAAPPPINKLALFPDKSAWCEAKNITQIVGHSGCEAKSIQNRACLGQCFSYSVPNTFPQSTESLVHCDSCMPAQSMWEIVTLECPGHEEVPRVDKLVEKILHCSCQACGKEPSHEGLSVYVQGEDGPGSQPGTHPHPHPHPHPGGQTPEPEDPPGAPHTEEEGAED
Especificaciones
AAH12037 | |
neuroblastoma, suppression of tumorigenicity 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NBL1 | |
GST |
4681 | |
Wheat germ expression system | |
25 μg | |
D1S1733E, DAN, DAND1, MGC8972, NB, NO3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |