missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAMPT/PBEF Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11776-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.Especificaciones
NAMPT/PBEF | |
Polyclonal | |
Unconjugated | |
NAMPT | |
1110035O14Rik, NAmPRTase, NAMPT, PBEF, PBEF1, Pre B cell enhancing factor, VF, VISFATIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100330160, 10135, 297508, 59027 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43490, Q80Z29, Q99KQ4 | |
Nampt, nampta | |
NAMPT/PBEF Fusion Protein Ag2434 | |
150 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
NAMPT/PBEF Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech > 150 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido