missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MYL2 (Human) Recombinant Protein
Human MYL2 full-length ORF ( AAH15821.1, 1 a.a. - 166 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00004633-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAPKKAKKRAGGANSNVFSMFEQTQIQEFKEAFTIMDQNRDGFIDKNDLRDTFAALGRVNVKNEEIDEMIKEAPGPINFTVFLTMFGEKLKGADPEETILNAFKVFDPEGKGVLKADYVREMLTTQAERFSKEEVDQMFAAFPPDVTGNLDYKNLVHIITHGEEKD
Especificaciones
AAH15821.1 | |
myosin, light chain 2, regulatory, cardiac, slow | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MYL2 | |
GST |
4633 | |
Wheat germ expression system | |
25 μg | |
CMH10, DKFZp779C0562, MLC2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |