missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myc Epitope Tag Antibody (2), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Bio-Techne NBP2-43627
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Myc Epitope Tag Monoclonal antibody specifically detects Myc Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Especificaciones
Myc Epitope Tag | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
Mouse | |
Protein G purified | |
RUO | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
2 | |
Western Blot 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:2000, Immunoprecipitation 1:100-1:500 | |
BHLHE39, Class E basic helix-loop-helix protein 39 (bHLHe39), Myc proto-oncogene protein, Proto-oncogene c-Myc, Transcription factor p64 | |
The immunogen used to generate this antibody corresponds to Myc tag .The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Purified | |
Epitope Tag |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Myc Epitope Tag Antibody (2), Novus Biologicals™ > 0.1mL; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido