Learn More
MP Biomedicals™ Trypsin, 1 - 250, From Hog Pancreas
From Hog Pancreas
Marca: MP Biomedicals™ 0210313980
Detalles adicionales : CAS : 9002-07-7 Peso : 0.10000kg
Descripción
Trypsin, 1 - 250, [9002-07-7]
- Parenzymol
- Dry powder: stable
- Protease act: min 250,000 USP units/gm
Dry Powder
Especificaciones
24000 | |
∽8 | |
Hog | |
Powder |
Trypsin | |
100g | |
4°C |