Learn More
MP Biomedicals™ Albumin, Bovine, RIA Grade
Prepared by heat shock fractionation
Marca: MP Biomedicals™ 0210503310
Detalles adicionales : CAS : 9048-46-8 Peso : 0.01000kg
Descripción
- Low fatty acids, IgG and hormones
- Purity: >98%
- pH: 7
- Protease-free
Mfr. No. 180537 may substitute
Especificaciones
ELISA, Western Blot | |
Bovine Serum Albumin | |
10g | |
4°C |
66.338 | |
6.8 to 7.2 | |
Bovine Serum | |
Crystalline |