Learn More
MP Biomedicals™ Albumin, Bovine, For IVD, Low IgG
Prepared by heat shock fractionation
Marca: MP Biomedicals™ 08810032
Detalles adicionales : CAS : 9048-46-8 Peso : 0.04950kg
Descripción
- For IVD applications, immunoassays, enzyme stabilization
- Low protease, conductivity and visible absorbance
- Purity: >98%
- pH: 7
Especificaciones
Immunoassay, Mammalian Cell Culture | |
Bovine Blood Albumin Fraction V | |
50g | |
0° to 5°C |
∽66 | |
7 | |
Bovine Blood | |
Crystalline |