Learn More
STEMCELL Technologies Mouse Recombinant IL-4
Interleukin 4
Marca:  STEMCELL Technologies 78047
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17179431
  • 797.85€ / 100 microgramos
Fecha estimada de envío: 20-05-2024
para ver el stock
Descripción
Descripción
  • Interleukin 4 (IL-4) is a type I cytokine produced by Th2 T cells, NK T cells, γ/δ T cells, eosinophils, mast cells, and activated basophils
  • IL-4 receptors are present on a number of cells, including hematopoietic, endothelial, epithelial, muscle, and brain tissues
  • IL-4 receptor engagement initiates signaling through the JAK/STAT pathway, and leads to the activation of PI3K and Ras/MAPK pathways (Nelms et al.) IL-4 plays a key role in immunoglobulin class switching in B cells and in regulating the differentiation of naïve T cells into T helper 2 (Th2) cells, while inhibiting Th1 differentiation
  • IL-4 acts together with tumor necrosis factor to stimulate expression of adhesion molecules on vascular endothelial cells and to downregulate expression of E-selectin, thus recruiting T cells and eosinophils to the site of inflammation (Nelms et al.)
Especificaciones
Especificaciones
B cell stimulating factor, BCDF, BCGF, BSF-1, Interleukin-4
100 μg
B Cells; Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; Monocytes; Myeloid Cells; Neurons; T Cells
≥95%