Learn More
STEMCELL Technologies Mouse Recombinant IL-22
Interleukin 22
Marca: STEMCELL Technologies 78039
Detalles adicionales : Peso : 0.00110kg
Descripción
- Interleukin 22 (IL-22) is a class 2 α-helical cytokine that signals through the class 2 cytokine receptor and activates the JAK/STAT pathway
- IL-22 is secreted by Th1, Th2, Tc22, and γδ T cells, dendritic, mast and NK cells
- It stimulates expression of antimicrobial peptides from epithelial cells, thus hindering bacterial infections
- Depending on the interactions with other cytokines, IL-22 can either promote inflammation or prevent tissue destruction by regulating host defense and tissue homeostasis at barrier surfaces (Sonnenberg et al.)
Especificaciones
IL-TIF, Interleukin-22 | |
100 μg | |
Airway Cells; Hepatic Cells; Intestinal Cells; Keratinocytes; T Cells; T Cells, CD4+; T Cells, CD8+ | |
≥95% |