Learn More
R&D Systems™ Mouse Mesenchymal Stem Cell Multi-Color Flow Kit Código de producto.: 18781864
R&D Systems™ Mouse Mesenchymal Stem Cell Multi-Color Flow Kit
Código de producto. 18781864
25 pruebas
Click to view available options
:
25 pruebas
Código de producto. 18781864
Marca: R&D Systems™ FMC003
For flow cytometric analysis of mouse MSCs using four fluorochrome-conjugated antibodies
Variations in techniques as well as differences in the MSC starting population may account for experimental variability and some of the contradictory data in the literature. By clearly defining the starting cell populations, researchers can decrease experimental variability and ultimately decrease apparent discrepancies in the literature that stem from the use of phenotypically different cell populations.
Verifies MSC identity in less than 2 hours
Included conjugated antibodies allow for single step staining
Defines the starting cell population
Multiple markers reduce experimental variation
TRUSTED_SUSTAINABILITY
Especificaciones
Contenido y almacenamiento Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Título del producto
R&D Systems™ Mouse Mesenchymal Stem Cell Multi-Color Flow Kit >