missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse immunoglobulin lambda light chain Rabbit anti-Mouse, Clone: RM110 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Marca: Abnova MAB14932.50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Rabbit monoclonal antibody raised against native mouse immunoglobulin lambda light chain.
- High specificity
- High reactivity
Especificaciones
Mouse immunoglobulin lambda light chain | |
Monoclonal | |
Biotin | |
ELISA (0.005-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blotting (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
P01843 | |
Igl-C1 | |
Native purified mouse IgM lambda. | |
50 μg | |
Primary | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
RM110 | |
Rabbit monoclonal antibody raised against native mouse immunoglobulin lambda light chain. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
Clambda1 | |
Rabbit | |
Protein A purification | |
RUO | |
110785 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mouse immunoglobulin lambda light chain Rabbit anti-Mouse, Clone: RM110 (Biotin), Abnova™ > 50ug
For Research Use Only (RUO)
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido