Learn More
ACROBiosystems Mouse Glypican 2 / GPC2 Protein, His Tag
399.17€ - 2896.79€
Especificaciones
Fuente Mouse Glypican 2, His Tag (GP2-M52H5) is expressed from human 293 cells (HEK293). It contains AA His 22 - Ser 556 (Accession # Q8BKV1-1).
Nombre común Glypican 2
Sistema de expresión HEK293
Formato Powder
Conjugado Unconjugated
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16467599
Ver documentos Detalles de la promoción
ACROBiosystems
GP2-M52H5-100UG
100 µg
399.17€
100 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 31-10-2022
para ver el stock
16477599
Ver documentos Detalles de la promoción
ACROBiosystems
GP2-M52H5-1MG
1 mg
2896.79€
1 miligramo
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 31-10-2022
para ver el stock
Descripción
Descripción
Glypican 2 (GPC2), also known cerebroglycan, is a glycophosphatidylinositol-linked integral membrane heparan sulfate proteoglycan found in the developing nervous system. Cerebroglycan participates in cell adhesion and is thought to regulate the growth and guidance of axons. Cerebroglycan has especially high affinity for laminin-1. GPC2 silencing inactivates Wnt/β-catenin signaling and reduces the expression of N-Myc, an oncogenic driver of neuroblastoma tumorigenesis. Immunotoxins and chimeric antigen receptor (CAR) T cells targeting GPC2 inhibit neuroblastoma growth in mouse models. A GPC3 specific antibody drug conjugate (ADC) can also inhibit neuroblastoma cell proliferation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.6 kDa. The protein migrates as 60-106 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
Mouse Glypican 2, His Tag (GP2-M52H5) is expressed from human 293 cells (HEK293). It contains AA His 22 - Ser 556 (Accession # Q8BKV1-1).
HEK293
Unconjugated
-20°C
Glypican 2
Powder
Bio-Markers and CD Antigens