Learn More
ACROBiosystems Mouse Glypican 1 / GPC1 Protein, His Tag
Marca: ACROBiosystems GP1-M52H7-1mg
Detalles adicionales : Peso : 0.00010kg
Descripción
Glypican 1 has been shown to interact with SLIT2. This protein is involved in the misfolding of normal prion proteins in the cell membrane to the infectious prion form. Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.5 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_057905.1 | |
57.5 kDa | |
1 mg | |
Asp 24 - Ser 529 | |
Glypican 1,GPC1 | |
Unconjugated | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Mouse |
Powder | |
>90% as determined by SDS-PAGE | |
Mouse Glypican 1, His Tag (GP1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 529 (Accession # Q9QZF2-1). | |
-20°C | |
Glypican 1 | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Bio-Markers and CD Antigens |