missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse Glypican 1 / GPC1 Protein, His Tag

419.88€ - 3083.12€

Especificaciones

Número de acceso NP_057905.1
Peso molecular 57.5 kDa
Fuente Mouse Glypican 1, His Tag (GP1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 529 (Accession # Q9QZF2-1).
Inmunógeno Asp 24 - Ser 529
Requisitos de almacenamiento -20°C
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16477589
Ver documentos Detalles de la promoción
ACROBiosystems
GP1-M52H7-1MG
1 mg
3083.12€
1 miligramo
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16467589
Ver documentos Detalles de la promoción
ACROBiosystems
GP1-M52H7-100UG
100 μg
419.88€
100 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Glypican 1 has been shown to interact with SLIT2. This protein is involved in the misfolding of normal prion proteins in the cell membrane to the infectious prion form. Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.5 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

NP_057905.1
Mouse Glypican 1, His Tag (GP1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 529 (Accession # Q9QZF2-1).
-20°C
Glypican 1
Mouse
50 mM Tris, 150 mM NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens
57.5 kDa
Asp 24 - Ser 529
Glypican 1,GPC1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Videos
SDS
Documentos

Documentos

Product Certifications