missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA6 Antibody, R&D Systems™
Rat Monoclonal Antibody
Marca: R&D Systems MAB6071
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
EphA6 Monoclonal specifically detects EphA6 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Especificaciones
EphA6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, Ehk2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, Hek12, PRO57066 | |
Rat | |
Protein A or G purified from ascites | |
RUO | |
285220 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Blockade of Receptor-ligand Interaction | |
81605 | |
Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
Q62413 | |
EPHA6 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA6 Met1-Gln546 Accession # Q62413 | |
500 μg | |
Primary | |
Detects mouse EphA6 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant rat EphB1 is observed and no cross-reactivity with recombinant human EphA1, A5, recombinant mouse EphA2, A3, A4, A7, A8, B2, B3, B4, or B6 is observed. | |
Mouse | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mouse EphA6 Antibody, R&D Systems™ > 500ug; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido