missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CXCL10 (IP-10) Recombinant Protein, PeproTech®
Click to view available options
Cantidad:
100 μg
1 mg
250 μg
25 μg
500 μg
50 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
25 microgramos
250 microgramos
5 microgramos
50 microgramos
500 microgramos
Descripción
Recombinant Murine IP-10 is an 8.7 kDa protein consisting of 77 amino acids, including the four conserved cysteine residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
Especificaciones
Especificaciones
| Número de acceso | P17515 |
| Para utilizar con (aplicación) | ELISA, Functional Assay, Western Blot |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 15945 |
| Peso molecular | 8.7 kDa |
| Nombre | Mouse CXCL10 (IP-10) |
| Cantidad | 25 μg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Gibco™ Mouse CXCL10 (IP-10) Recombinant Protein, PeproTech® >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido

