missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD30 Ligand/TNFSF8 Antibody, R&D Systems™
Goat Polyclonal Antibody
Marca: R&D Systems AF732
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD30 Ligand/TNFSF8 Polyclonal specifically detects CD30 Ligand/TNFSF8 in Mouse samples. It is validated for Western Blot, Neutralization.
Especificaciones
CD30 Ligand/TNFSF8 | |
Polyclonal | |
Unconjugated | |
P32972 | |
TNFSF8 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD30 Ligand/TNFSF8 Gln68-Asp239 Accession # P32972 | |
100 μg | |
Primary | |
Detects mouse CD30 Ligand/TNFSF8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human CD30 Ligand is observed. | |
Mouse | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 3-10 ug/mL | |
CD153, CD153 antigen, CD30 antigen ligand, CD30L, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, TNFSF8, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8 | |
Goat | |
Affinity Purified | |
RUO | |
944 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mouse CD30 Ligand/TNFSF8 Antibody, R&D Systems™ > 100μg; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido