Learn More
Hepatitis E virus ORF 3, Mouse, Clone: 5576f, Abnova™
Mouse monoclonal antibody raised against partial recombinant Hepatitis E virus ORF 3.
Marca: Abnova MAB8940.100uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Hepatitis E virus ORF 3 | |
Monoclonal | |
1 ug/uL | |
Mouse monoclonal antibody raised against partial recombinant Hepatitis E virus ORF 3. | |
In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300) | |
Recombinant protein corresponding to amino acids 34-123 of Hepatitis E virus ORF 3. | |
100 μL | |
Primary | |
Virus | |
Liquid |
ELISA, Western Blot | |
5576f | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Mouse | |
Affinity purification | |
RUO | |
This antibody recognizes recombinant protein Hepatitis E virus ORF 3. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Título del producto
Hepatitis E virus ORF 3, Mouse, Clone: 5576f, Abnova™ > 100μL; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido