Learn More
HGF Mouse anti-Human, Clone: 24612, R&D Systems™

Mouse Monoclonal Antibody has been used in 15 publications
Marca: R&D Systems MAB294-500

Descripción
HGF Monoclonal specifically detects HGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Especificaciones
HGF | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3082 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Neutralization | |
24612 | |
Western Blot 1 ug/mL, Neutralization 0.1-0.3 ug/mL | |
P14210 | |
HGF | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HGF Gln32-Ser728 Accession # P14210 | |
500 μg | |
Primary | |
Detects human HGF in Western blots. In Western blots, no-cross reactivity with recombinant mouse HGF or recombinant canine HGF is observed. | |
Human | |
Supernatant |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.