Learn More
CGA, Mouse, Clone: ME.132, Abnova™
Mouse monoclonal antibody raised against CGA.
Marca: Abnova MAB4749.5mg
Descripción
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family. [provided by RefSeq
Especificaciones
CGA | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
CGA | |
CGA | |
Human CGA. | |
RUO | |
1081 | |
Human | |
Liquid |
ELISA | |
ME.132 | |
Mouse monoclonal antibody raised against CGA. | |
In PBS, pH 7.4 | |
CG-ALPHA/FSHA/GPHA1/GPHa/HCG/LHA/TSHA | |
Mouse | |
5 mg | |
Primary | |
human TSH-alpha, MW 28kD. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.