Learn More
CACNA1H, Mouse, Clone: S55-10, Abnova™
Mouse monoclonal antibody raised against partial recombinant CACNA1H.
Marca: Abnova MAB6650.100ug
Descripción
This gene encodes a T-type member of the alpha-1 subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. The alpha-1 subunit has 24 transmembrane segments and forms the pore through which ions pass into the cell. There are multiple isoforms of each of the proteins in the complex, either encoded by different genes or the result of alternative splicing of transcripts. Alternate transcriptional splice variants, encoding different isoforms, have been characterized for the gene described here. Studies suggest certain mutations in this gene lead to childhood absence epilepsy (CAE). [provided by RefSeq
Especificaciones
CACNA1H | |
Monoclonal | |
Unconjugated | |
Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1.0-10 ug/mL) The optimal working dilution should be determined by the end user. | |
CACNA1H | |
CACNA1H | |
Recombinant fusion protein corresponding to amino acids 1019-1293 (II-III loop) of human CACNA1H. | |
RUO | |
8912 | |
Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
S55-10 | |
Mouse monoclonal antibody raised against partial recombinant CACNA1H. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
CACNA1HB/Cav3.2/EIG6/FLJ90484 | |
Mouse | |
100 μg | |
Primary | |
Detects ∼260KDa. No cross-reactivity against Cav1.3. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.