Learn More
Cyclin D3 Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry applications.
Marke: Epredia MS-215-R7
Beschreibung
Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. Cyclins D1, D2, and D3 form active kinase complexes in vivo with a distinct subset of cdks, particularly with cdk4 and cdk6.Host Species: Mouse
Clone: DCS-22
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified human recombinant full-length cyclin D3 protein
Molecular Weight: 34 kDa
Positive Control: LS174T cells, Breast CA or tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native only)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO

Spezifikation
Cyclin D3 Ab-1 | |
Monoclonal | |
— | |
Mouse | |
7 mL | |
Cell Cycle | |
Human, Mouse, Non-human Primate, Rat | |
IgG2a κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
DCS-22 | |
Unconjugated | |
Purified human recombinant full length cyclin D3 protein | |
RUO | |
Primary | |
Prediluted |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.