Learn More
JMJD2A Recombinant Rabbit Monoclonal Antibody (6H56L3), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen 702449
404.26 EUR hasta el 2025-06-30
Use el código promocional "24108" para aplicar la oferta.
Descripción
This antibody is predicted to react with Monkey, Cat, and Horse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
JMJD2A is a member of the Jumonji domain 2 (JMJD2) family and a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
Especificaciones
JMJD2A | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O75164 | |
KDM4A | |
Protein corresponding to human (aa379-aa628) | |
100μg | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay, Immunocytochemistry | |
6H56L3 | |
Unconjugated | |
KDM4A | |
D4Ertd222e; JHDM3A; jmjC domain-containing histone demethylation protein 3A; JMJD2; JMJD2A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; jumonji domain-containing protein 2A; KDM4A; KIAA0677; lysine (K)-specific demethylase 4A; lysine demethylase 4A; lysine-specific demethylase 4A; mKIAA0677; TDRD14A; tudor domain containing 14A | |
Rabbit | |
Protein A | |
RUO | |
9682 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.