missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human JAK1, GST Tag Recombinant Protein
Janus kinase 1 (JAK1) is a protein tyrosine kinase.
Marca: Invitrogen™ PV4775
Detalles adicionales : Peso : 0.01000kg
Descripción
JAK1 is involved in the interferon-alpha/beta and -gamma signal transduction pathways.
Especificaciones
NP_002218.2 | |
1mg | |
JAK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
60.8 kDa | |
JAK1, JAK1A, JAK1B | |
Human | |
GST Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Cytoplasmic, Catalytic | |
Signal Transduction |