missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL3 (Human) Recombinant Protein
Human IL3 (149 a.a.) full-lenth recombinant protein with His tag expressed in Barley grain (Hordeum vulgare). This protein is endotoxins-free and thus optimal to use in stem cell cultures.
Marca: Abnova™ P5884.100ug
Detalles adicionales : Peso : 0.00010kg
Especificaciones
3562 | |
Hordeum vulgare (barley) expression system | |
100 μg | |
IL-3, MCGF, MGC79398, MGC79399, MULTI-CSF | |
Human | |
His | |
Lyophilized |
interleukin 3 (colony-stimulating factor, multiple) | |
SDS-PAGE Stained with Coomassie Blue | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
IL3 | |
Plants | |
Lyophilized from PBS, pH 7.2 |