missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK beta Recombinant Rabbit Monoclonal Antibody (5D12), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA549817
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Phosphorylation of serine residues on the I-kappa-B proteins by IKK kinases, including IKBKB, marks them for destruction via the ubiquitination pathway, allowing activation of the NF-kappa-B complex. Inhibitor of nuclear factor kappa-B kinase subunit beta (IKK beta) is a serine kinase that plays an essential role in the NF-kaapa-B signaling pathway which is activated by mulitple stimuli such as inflammatory cytokines, bacterial, or viral products, DNA damages, or other cellular stresses. IKK beta acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Mutations affecting the IKK beta gene can result in Immunodeficiency 15 (IMD15).
Especificaciones
IKK beta | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.4 | |
O14920 | |
Ikbkb | |
A synthesized peptide derived from human IKK beta | |
100 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
5D12 | |
Unconjugated | |
Ikbkb | |
AI132552, AIM-1, FLJ40509, I-kappa-B kinase 2, I-kappa-B-kinase beta, Ikbkb, IKK[b], IKK2, IKK-2, IKKB, IKK-B, IKKbeta, IKK-beta, IMD15, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of kappaB kinase beta, inhibitor of nuclear factor kappa B kinase beta subunit, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKB, nuclear factor NF-kappa-B inhibitor kinase beta | |
Rabbit | |
Affinity chromatography | |
RUO | |
3551 | |
-20°C or -80°C if preferred | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
IKK beta Recombinant Rabbit Monoclonal Antibody (5D12), Invitrogen™ > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido