missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IKBKE (IKK Epsilon), GST Tag Recombinant Protein
Human IKBKE (IKK Epsilon), GST Tag Recombinant Protein
Marca: Invitrogen™ PV4876
Detalles adicionales : Peso : 0.01000kg
Descripción
Over expression of IKBKE has been observed in breast cancer cells, and suppression of IKBKE induces cell death.
Especificaciones
NP_054721 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Polysorbate 20, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
IKBKE (IKK Epsilon) | |
Human | |
GST Tag | |
Frozen |
Kinase Assay, Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
107.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
IKK Kinase Family | |
Recombinant | |
Baculovirus |