Learn More
IgG1 Mouse, FITC, Clone: MOPC-21, Isotype Control, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60070FI
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17128841
  • 203.85€ / 200 microgramos
Explore más ofertas especiales
Descripción
Descripción
  • The MOPC-21 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
  • Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
  • If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
  • The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
  • This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Especificaciones
Especificaciones
IgG1
Monoclonal
FITC
200 μg
IgG1 κ
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
MOPC-21
Mouse
Isotype Control (Primary)
Other
Certificados
Ofertas especiales
Ofertas especiales
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
IgG1 Mouse, FITC, Clone: MOPC-21, Isotype Control, STEMCELL Technologies