missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium tuberculosis Rabbit anti-Bacteria, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Marca: Abnova PAB29798.500uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Rabbit polyclonal antibody raised against Mycobacterium tuberculosis.
- High specificity
- High reactivity
Especificaciones
Mycobacterium tuberculosis | |
Polyclonal | |
Rabbit polyclonal antibody raised against Mycobacterium tuberculosis. | |
In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide). | |
Native purified protein derivative from Mycobacterium tuberculosis. | |
RUO | |
This antibody reacts with Mycobacterium tuberculosis. Reactive with several related microbes including M. avium, M. phlei, and M. parafortuitum. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow, Western Blot | |
Unconjugated | |
ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user. | |
Rabbit | |
500 μL | |
Primary | |
Bacteria | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mycobacterium tuberculosis Rabbit anti-Bacteria, Polyclonal, Abnova™ > 500uL
For Research Use Only (RUO)
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido