Learn More
Abnova™ Human WASF1 Partial ORF (NP_003922.1, 198 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008936-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene, a member of the Wiskott-Aldrich syndrome protein (WASP)-family, plays a critical role downstream of Rac, a Rho-family small GTPase, in regulating the actin cytoskeleton required for membrane ruffling. It has been shown to associate with an actin nucleation core Arp2/3 complex while enhancing actin polymerization in vitro. Wiskott-Aldrich syndrome is a disease of the immune system, likely due to defects in regulation of actin cytoskeleton. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: HDRRREWQKLAQGPELAEDDANLLHKHIEVANGPASHFETRPQTYVDHMDGSYSLSALPFSQMSELLTRAEERVLVRPHEPPPPPPMHGAGDAKPIPTCIEspecificaciones
NP_003922.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HDRRREWQKLAQGPELAEDDANLLHKHIEVANGPASHFETRPQTYVDHMDGSYSLSALPFSQMSELLTRAEERVLVRPHEPPPPPPMHGAGDAKPIPTCI | |
RUO | |
WASF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8936 | |
WASF1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31482/KIAA0269/SCAR1/WAVE/WAVE1 | |
WASF1 | |
Recombinant | |
wheat germ expression system |