missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human V1a Vasopressin R/AVPR1A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB10932-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
V1a Vasopressin R/AVPR1A Monoclonal antibody specifically detects V1a Vasopressin R/AVPR1A in Human samples. It is validated for Flow Cytometry
Especificaciones
V1a Vasopressin R/AVPR1A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
Antidiuretic hormone receptor 1a, arginine vasopressin receptor 1A, AVPR V1a, AVPR1, AVPR1A, SCCL vasopressin subtype 1a receptor, V1a vasopressin receptor, V1a VasopressinR, V1aR, V1-vascular vasopressin receptor AVPR1A, Vascular/hepatic-type arginine vasopressin receptor, vasopressin V1a receptor | |
NS0 mouse myeloma cell line transfected with human V1a Vasopressin R/AVPR1A, Accession # P37288 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry | |
612130 | |
Flow Cytometry 0.25 ug/10^6 cells | |
P37288 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
552 | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human V1a Vasopressin R/AVPR1A Antibody, R&D Systems™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido