Learn More
Abnova™ Human UXT Partial ORF (NP_705582, 10 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008409-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a novel protein which is highly conserved in mouse. It interacts with the N-terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is also likely to be involved in tumorigenesis as it is abundantly expressed in tumor tissues. This gene is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: EPIMATPPKRRAVEATGEKVLRYETFISDVLQRDLRKVLDHRDKVYEQLAKYLQLRNVIERLQEAKHSELYMQVDLGCNFFVDTVVPDTSREspecificaciones
NP_705582 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EPIMATPPKRRAVEATGEKVLRYETFISDVLQRDLRKVLDHRDKVYEQLAKYLQLRNVIERLQEAKHSELYMQVDLGCNFFVDTVVPDTSR | |
RUO | |
UXT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8409 | |
UXT (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ART-27 | |
UXT | |
Recombinant | |
wheat germ expression system |