Learn More
Abnova™ Human USP11 Partial ORF (NP_004642, 201 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008237-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Protein ubiquitination controls many intracellular processes, including cell cycle progression, transcriptional activation, and signal transduction. This dynamic process, involving ubiquitin conjugating enzymes and deubiquitinating enzymes, adds and removes ubiquitin. Deubiquitinating enzymes are cysteine proteases that specifically cleave ubiquitin from ubiquitin-conjugated protein substrates. This gene encodes a deubiquitinating enzyme which lies in a gene cluster on chromosome Xp11.23 [provided by RefSeq]
Sequence: LVRHNDLGKSHTVQFSHTDSIGLVLRTARERFLVEPQEDTRLWAKNSEGSLDRLYDTHITVLDAALETGQLIIMETRKKDGTWPSAQLHVMNNNMSEEDEEspecificaciones
NP_004642 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LVRHNDLGKSHTVQFSHTDSIGLVLRTARERFLVEPQEDTRLWAKNSEGSLDRLYDTHITVLDAALETGQLIIMETRKKDGTWPSAQLHVMNNNMSEEDE | |
RUO | |
USP11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8237 | |
USP11 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UHX1 | |
USP11 | |
Recombinant | |
wheat germ expression system |