Learn More
Abnova™ Human TRAF7 Partial ORF (AAH24267, 61 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084231-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Tumor necrosis factor (TNF; see MIM 191160) receptor-associated factors, such as TRAF7, are signal transducers for members of the TNF receptor superfamily (see MIM 191190). TRAFs are composed of an N-terminal cysteine/histidine-rich region containing zinc RING and/or zinc finger motifs; a coiled-coil (leucine zipper) motif; and a homologous region that defines the TRAF family, the TRAF domain, which is involved in self-association and receptor binding.[supplied by OMIM]
Sequence: VFKDPVITTCGHTFCRRCALKSEKCPVDNVKLTVVVNNIAVAEQIGELFIHCRHGCRVAGSGKPPIFEVDPRGCPFTIKLSARKDHEGSCDYRPVRCPNNEspecificaciones
AAH24267 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VFKDPVITTCGHTFCRRCALKSEKCPVDNVKLTVVVNNIAVAEQIGELFIHCRHGCRVAGSGKPPIFEVDPRGCPFTIKLSARKDHEGSCDYRPVRCPNN | |
RUO | |
TRAF7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84231 | |
TRAF7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586I021/MGC7807/RFWD1/RNF119 | |
TRAF7 | |
Recombinant | |
wheat germ expression system |