Learn More
Abnova™ Human TMPRSS5 Partial ORF (NP_110397.1, 258 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080975-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that belongs to the serine protease family. Serine proteases are known to be involved in many physiological and pathological processes. [provided by RefSeq]
Sequence: HCMHSFRLARLSSWRVHAGLVSHSAVRPHQGALVERIIPHPLYSAQNHDYDVALLRLQTALNFSDTVGAVCLPAKEQHFPKGSRCWVSGWGHTHPSHTYSEspecificaciones
NP_110397.1 | |
Liquid | |
80975 | |
TMPRSS5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC141886/MGC148044/SPINESIN | |
TMPRSS5 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HCMHSFRLARLSSWRVHAGLVSHSAVRPHQGALVERIIPHPLYSAQNHDYDVALLRLQTALNFSDTVGAVCLPAKEQHFPKGSRCWVSGWGHTHPSHTYS | |
RUO | |
TMPRSS5 | |
Wheat Germ (in vitro) | |
GST |