Learn More
Abnova™ Human SYT13 Partial ORF (NP_065877.1, 36 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057586-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
SYT13 belongs to the large synaptotagmin protein family. All synaptotagmins show type I membrane topology, with an extracellular N terminus, a single transmembrane region, and a cytoplasmic C terminus containing tandem C2 domains. Major functions of synaptotagmins include vesicular traffic, exocytosis, and secretion.[supplied by OMIM]
Sequence: KKGLLPRDQDPDLEKAKPSLLGSAQQFNVKKSTEPVQPRALLKFPDIYGPRPAVTAPEVINYADYSLRSTEEPTAPASPQPPNDSRLKRQVTEELFILPQEspecificaciones
NP_065877.1 | |
Liquid | |
57586 | |
SYT13 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1427 | |
SYT13 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KKGLLPRDQDPDLEKAKPSLLGSAQQFNVKKSTEPVQPRALLKFPDIYGPRPAVTAPEVINYADYSLRSTEEPTAPASPQPPNDSRLKRQVTEELFILPQ | |
RUO | |
SYT13 | |
Wheat Germ (in vitro) | |
GST |