Learn More
Abnova™ Human ST8SIA1 Partial ORF (NP_003025, 61 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006489-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Gangliosides are membrane-bound glycosphingolipids containing sialic acid. Ganglioside GD3 is known to be important for cell adhesion and growth of cultured malignant cells. The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to GM3 to produce gangliosides GD3 and GT3. The encoded protein may be found in the Golgi apparatus and is a member of glycosyltransferase family 29. [provided by RefSeq]
Sequence: VLQQGTAWRRNQTAARAFRKQMEDCCDPAHLFAMTKMNSPMGKSMWYDGEFLYSFTIDNSTYSLFPQATPFQLPLKKCAVVGNGGILKKSGCGRQIDEANSpecifica
NP_003025 | |
Liquid | |
6489 | |
ST8SIA1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GD3S/SIAT8/SIAT8A/ST8SiaI | |
ST8SIA1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VLQQGTAWRRNQTAARAFRKQMEDCCDPAHLFAMTKMNSPMGKSMWYDGEFLYSFTIDNSTYSLFPQATPFQLPLKKCAVVGNGGILKKSGCGRQIDEAN | |
RUO | |
ST8SIA1 | |
Wheat Germ (in vitro) | |
GST |