missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST8SIA1 Partial ORF (NP_003025, 61 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Especificaciones

Número de acceso NP_003025
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 6489
Peso molecular 36.74kDa
Ver más especificaciones

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
16155295
View Documents Promotion Details
Abnova™
H00006489-Q01.10UG
10 ug
335.00€
10 microgramos
View alternative products
Save Up to 
»
Fecha estimada de envío: 07-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16165295
View Documents Promotion Details
Abnova™
H00006489-Q01.25UG
25 ug
508.00€
25 microgramos
View alternative products
Save Up to 
»
Fecha estimada de envío: 07-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

Gangliosides are membrane-bound glycosphingolipids containing sialic acid. Ganglioside GD3 is known to be important for cell adhesion and growth of cultured malignant cells. The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to GM3 to produce gangliosides GD3 and GT3. The encoded protein may be found in the Golgi apparatus and is a member of glycosyltransferase family 29. [provided by RefSeq]

Sequence: VLQQGTAWRRNQTAARAFRKQMEDCCDPAHLFAMTKMNSPMGKSMWYDGEFLYSFTIDNSTYSLFPQATPFQLPLKKCAVVGNGGILKKSGCGRQIDEAN
Specifications

Specifications

NP_003025
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GD3S/SIAT8/SIAT8A/ST8SiaI
ST8SIA1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
6489
ST8SIA1 (Human) Recombinant Protein (Q01)
VLQQGTAWRRNQTAARAFRKQMEDCCDPAHLFAMTKMNSPMGKSMWYDGEFLYSFTIDNSTYSLFPQATPFQLPLKKCAVVGNGGILKKSGCGRQIDEAN
RUO
ST8SIA1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications