missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST3GAL1 Partial ORF (NP_003024, 29 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006482-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16165275

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 12-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of the encoded protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B. Two transcript variants encoding the same protein have been found for this gene. Other transcript variants may exist, but have not been fully characterized yet. [provided by RefSeq]

Sequence: SHTMVATTWFPKQMVLELSENLKRLIKHRPCTCTHCIGQRKLSAWFDERFNQTMQPLLTAQNALLEDDTYRWWLRLQREKKPNNLNDTIKELFRVVPGNV
Especificaciones

Especificaciones

NP_003024
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SHTMVATTWFPKQMVLELSENLKRLIKHRPCTCTHCIGQRKLSAWFDERFNQTMQPLLTAQNALLEDDTYRWWLRLQREKKPNNLNDTIKELFRVVPGNV
RUO
ST3GAL1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6482
ST3GAL1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp666E036/DKFZp779K2051/FLJ36548/Gal-NAc6S/MGC9183/SIAT4A/SIATFL/ST3GalA/ST3GalA.1/ST3GalIA/ST3GalIA,1/ST3O
ST3GAL1
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales