Learn More
Abnova™ Human SLC35B2 Partial ORF (NP_835361.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00347734-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sulfotransferases (e.g., SULT4A1; MIM 608359) use an activated form of sulfate, 3-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS), as a common sulfate donor for sulfation of glycoproteins, proteoglycans, and glycolipids in the endoplasmic reticulum and Golgi apparatus. SLC35B2 is located in the microsomal membrane and transports PAPS from the cytosol, where it is synthesized, into the Golgi lumen (Kamiyama et al., 2003 [PubMed 12716889]).[supplied by OMIM]
Sequence: MDARWWAVVVLAAFPSLGAGGETPEAPPESWTQLWFFRFVVNAAGYASFMVPGYLLVQYFRRKNYLETGRGLCFPLVKACVFGNEPKASDEVPLAPRTEAEspecificaciones
NP_835361.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MDARWWAVVVLAAFPSLGAGGETPEAPPESWTQLWFFRFVVNAAGYASFMVPGYLLVQYFRRKNYLETGRGLCFPLVKACVFGNEPKASDEVPLAPRTEA | |
RUO | |
SLC35B2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
347734 | |
SLC35B2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAPST1/SLL/UGTrel4 | |
SLC35B2 | |
Recombinant | |
wheat germ expression system |