Learn More
ACROBiosystems Human Siglec-3 / CD33 Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
Marca:  ACROBiosystems CD3-H5259-1MG
Detalles adicionales : Peso : 0.00001kg
 Ver más versiones de este producto
Código de producto. 15806177
  • 2818.49€ / 1 miligramo
Fecha estimada de envío: 29-05-2024
para ver el stock
Descripción
Descripción
Myeloid cell surface antigen CD33 is also known as SIGLEC3, Siglecs (sialic acid binding Ig­like lectins) and GP67, is a single-pass type I membrane protein which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. Human CD33 / Siglec-3 cDNA encodes a 364 amino acid (aa) polypeptide with a hydrophobic signal peptide, an N-­terminal Ig-­like V­-type domain, one Ig­-like C2-­type domains, a transmembrane region and a cytoplasmic tail. CD33 / Siglec-3 usually considered myeloid-specific, but it can also be found on some lymphoid cells. In the immune response, CD33 / Siglec-3 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. CD33 / Siglec-3 induces apoptosis in acute myeloid leukemia.
This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 54.6 kDa. The protein migrates as 64-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
AAH28152.1
1 mg
Asp 18 - His 259
CD33,SIGLEC3,gp67
Unconjugated
Llama IgG Fc Tag
Less than 0.01 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
54.6 kDa
Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin (CD3-H5259) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).
-20°C
Siglec-3
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens